Learn More
Biomatik Corporation Mouse Glutamate Receptor, Ionotropic, AMPA 4 (GRIA4) ELISA Kit, 96 tests
Catalog No. 501502608
Encompass_Preferred
Mouse Glutamate Receptor, Ionotropic, AMPA 4 (GRIA4) ELISA Kit for detecting GRIA4 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Z2W8. Gene ID: 14802. Target Synonyms: GLUR4; GLUR4C; GLURD; AMPA-selective glutamate receptor 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2608 Supplier Biomatik Corporation Supplier No. EKU04453
Mouse Glutamate Receptor, Ionotropic, AMPA 4 (GRIA4) ELISA Kit for detecting GRIA4 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Z2W8. Gene ID: 14802. Target Synonyms: GLUR4; GLUR4C; GLURD; AMPA-selective glutamate receptor 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.