Learn More
Biomatik Corporation Mouse Glutaminase (GLS) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04462
Encompass_Preferred
Mouse Glutaminase (GLS) ELISA Kit for detecting GLS in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.118ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: D3Z7P3. Gene ID: 14660. Target Synonyms: GLS1; Glutaminase kidney isoform, mitochondrial; K-glutaminase; L-glutamine amidohydrolase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2610