Learn More
Biomatik Corporation Human Lacritin (LACRT) ELISA Kit
Supplier:  Biomatik Corporation EKU05526
Encompass_Preferred
Human Lacritin (LACRT) ELISA Kit for detecting LACRT in saliva, tears, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.63ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9GZZ8. Gene ID: 90070. Target Synonyms: Extracellular glycoprotein lacritin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6172