Learn More
Biomatik Corporation Human Lactoperoxidase (LPO) ELISA Kit
Supplier:  Biomatik Corporation EKU05544
Encompass_Preferred
Human Lactoperoxidase (LPO) ELISA Kit for detecting LPO in breast milk, tear, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000ng/ml. Sensitivity: 6.1ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P22079. Gene ID: 4025. Target Synonyms: SPO; Salivary peroxidase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6178