Learn More
Biomatik Corporation Dog Leptin (LEP) ELISA Kit
Supplier:  Biomatik Corporation EKU05590
Encompass_Preferred
Dog Leptin (LEP) ELISA Kit for detecting LEP in serum, plasma, other biological fluids. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 0.312-20ng/ml. Sensitivity: 0.112ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O02720. Gene ID: 403616. Target Synonyms: OB; OBS; Obesity Homolog; Obesity Factor; Obese Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6183