Learn More
Biomatik Corporation Human Glutaredoxin (GLRX) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04470
Encompass_Preferred
The Human Glutaredoxin (GLRX) ELISA Kit allows for quantitative determination of Glutaredoxin in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.94-60 ng/mL. Sensitivity: 0.41 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P35754. Cross Activity: No significant cross-reactivity or interference between Glutaredoxin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2614