Learn More
Biomatik Corporation Human Glutathione Peroxidase 1 (GPX1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04475
Encompass_Preferred
The Human Glutathione Peroxidase 1 (GPX1) ELISA Kit allows for quantitative determination of Glutathione Peroxidase 1 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 3.12-200 ng/mL. Sensitivity: 5.2 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P07203. Cross Activity: No significant cross-reactivity or interference between Glutathione Peroxidase 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2618