Learn More
Biomatik Corporation Mouse Glutathione Peroxidase 1 (GPX1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04476
Encompass_Preferred
Mouse Glutathione Peroxidase 1 (GPX1) ELISA Kit for detecting GPX1 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.117ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P11352. Gene ID: 14775. Target Synonyms: GSHPX1; Cellular glutathione peroxidase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2619