Learn More
Biomatik Corporation Human Glutathione Peroxidase 2, Gastrointestinal (GPX2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04479
Encompass_Preferred
The Human Glutathione Peroxidase 2, Gastrointestinal (GPX2) ELISA Kit allows for quantitative determination of Glutathione Peroxidase 2, Gastrointestinal in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.116 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P18283. Cross Activity: No significant cross-reactivity or interference between Glutathione Peroxidase 2, Gastrointestinal and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2621