Learn More
Biomatik Corporation Bovine Matrix Metalloproteinase 9 (MMP9) ELISA Kit
Supplier:  Biomatik Corporation EKU05878
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenatesand other biological fluids. Species Reactivity: Bovine. Detection Range: 0.312-20ng/mL. Sensitivity: 0.123ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P52176. Target Synonyms: GELB; Gelatinase B; CLG4B; CLG4-B; 92 KDa Gelatinase; 92kDa Type IV Collagenase. Restriction: For Research Use Only.
Catalog No. 50-228-6216