Learn More
Biomatik Corporation Human Glutathione Peroxidase 4 (GPX4) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04482
Encompass_Preferred
The Human Glutathione Peroxidase 4 (GPX4) ELISA Kit allows for quantitative determination of Glutathione Peroxidase 4 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 3.12-200 ng/mL. Sensitivity: 1.17 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q4AEH2. Cross Activity: No significant cross-reactivity or interference between Glutathione Peroxidase 4 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2623