Learn More
Biomatik Corporation Human Glutathione Reductase (GR) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04483
Encompass_Preferred
The Human Glutathione Reductase (GR) ELISA Kit allows for quantitative determination of Glutathione Reductase in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 78-5,000 pg/mL. Sensitivity: 35 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P00390. Cross Activity: No significant cross-reactivity or interference between Glutathione Reductase and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2624