Learn More
Biomatik Corporation Human Glutathione S Transferase Alpha 1 (GSTa1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04486
Encompass_Preferred
The Human Glutathione S Transferase Alpha 1 (GSTa1) ELISA Kit allows for quantitative determination of Glutathione S Transferase Alpha 1 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 62.5-4,000 pg/mL. Sensitivity: 27.4 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P08263. Cross Activity: No significant cross-reactivity or interference between Glutathione S Transferase Alpha 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2625