Learn More
Biomatik Corporation Human Glutathione S Transferase Alpha 2 (GSTa2) ELISA Kit, 96 tests
Catalog No. 501502626
Encompass_Preferred
The Human Glutathione S Transferase Alpha 2 (GSTa2) ELISA Kit allows for quantitative determination of Glutathione S Transferase Alpha 2 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.125 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P09210. Cross Activity: No significant cross-reactivity or interference between Glutathione S Transferase Alpha 2 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2626 Supplier Biomatik Corporation Supplier No. EKU04489
The Human Glutathione S Transferase Alpha 2 (GSTa2) ELISA Kit allows for quantitative determination of Glutathione S Transferase Alpha 2 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.125 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P09210. Cross Activity: No significant cross-reactivity or interference between Glutathione S Transferase Alpha 2 and analogues was observed. Restriction: For Research Use Only.