Learn More
Biomatik Corporation Human Glutathione S Transferase Pi (GSTp) ELISA Kit, 96 tests
Catalog No. 501502627
Encompass_Preferred
The Human Glutathione S Transferase Pi (GSTp) ELISA Kit allows for quantitative determination of Glutathione S Transferase Pi in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.056 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P09211. Cross Activity: No significant cross-reactivity or interference between Glutathione S Transferase Pi and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2627 Supplier Biomatik Corporation Supplier No. EKU04497
The Human Glutathione S Transferase Pi (GSTp) ELISA Kit allows for quantitative determination of Glutathione S Transferase Pi in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.056 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P09211. Cross Activity: No significant cross-reactivity or interference between Glutathione S Transferase Pi and analogues was observed. Restriction: For Research Use Only.