Learn More
Biomatik Corporation Mouse Glyceraldehyde-3-Phosphate Dehydrogenase (GAPDH) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04505
Encompass_Preferred
Mouse Glyceraldehyde-3-Phosphate Dehydrogenase (GAPDH) ELISA Kit for detecting GAPDH in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P16858. Gene ID: 14433. Target Synonyms: G3PD; GAPD; Peptidyl-cysteine S-nitrosylase GAPDH. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2630