Learn More
Biomatik Corporation Rat Myeloid Differentiation Factor 88 (MyD88) ELISA Kit
Supplier:  Biomatik Corporation EKU06061
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Rat. Detection Range: 78-5,000pg/mL. Sensitivity: 30pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q6Y1S1. Target Synonyms: Myeloid Differentiation Primary Response Gene 88. Restriction: For Research Use Only.
Catalog No. 50-228-6241