Learn More
Biomatik Corporation Chicken Myostatin (MSTN) ELISA Kit
Supplier:  Biomatik Corporation EKU06114
Encompass_Preferred
Chicken Myostatin (MSTN) ELISA Kit for detecting MSTN in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.224ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O42220. Gene ID: 373964. Target Synonyms: GDF8; Growth Differentiation Factor 8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6247