Learn More
Biomatik Corporation Rat Glycoprotein 39, Cartilage (GP39) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04528
Encompass_Preferred
Rat Chitinase-3-like Protein 1 (CHI3L1) ELISA Kit for detecting CHI3L1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 3.12-200ng/ml. Sensitivity: 1.27ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9WTV1. Gene ID: 89824. Target Synonyms: GP39; CHI3-L1; YKL40; Chondrex; Chitinase 3-Like 1; Chondrocyte Protein YKL40; 39 kDa synovial protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2635