Learn More
Biomatik Corporation Human Neurofilament 3 (NEF3) ELISA Kit
Supplier:  Biomatik Corporation EKU06212
Encompass_Preferred
Human Neurofilament 3 (NEF3) ELISA Kit for detecting NEF3 in serum, plasma, tissue homogenates, cerebrospinal fluid, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P07197. Gene ID: 4741. Target Synonyms: NEFM; NF-M; NFM; Neurofilament,Medium Polypeptide; 160 kDa neurofilament protein; Neurofilament triplet M protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6254