Learn More
Biomatik Corporation Human Glycoprotein VI, Platelet (GP6) ELISA Kit, 96 tests
Catalog No. 501502637
Encompass_Preferred
The Human Glycoprotein VI, Platelet (GP6) ELISA Kit allows for quantitative determination of Glycoprotein VI, Platelet in Plasma. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1000 pg/mL. Sensitivity: 6.3 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9HCN6. Cross Activity: No significant cross-reactivity or interference between Glycoprotein VI, Platelet and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2637 Supplier Biomatik Corporation Supplier No. EKU04534
The Human Glycoprotein VI, Platelet (GP6) ELISA Kit allows for quantitative determination of Glycoprotein VI, Platelet in Plasma. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1000 pg/mL. Sensitivity: 6.3 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9HCN6. Cross Activity: No significant cross-reactivity or interference between Glycoprotein VI, Platelet and analogues was observed. Restriction: For Research Use Only.