Learn More
Biomatik Corporation Human Glyoxalase I (GLO1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04539
Encompass_Preferred
The Human Glyoxalase I (GLO1) ELISA Kit allows for quantitative determination of Glyoxalase I in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.59 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q04760. Cross Activity: No significant cross-reactivity or interference between Glyoxalase I and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2639