Learn More
Biomatik Corporation Rat Nitric Oxide Synthase 1, Neuronal (NOS1) ELISA Kit
Supplier:  Biomatik Corporation EKU06306
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Rat. Detection Range: 0.156-10ng/mL. Sensitivity: 0.055ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P29476. Target Synonyms: NOS; nNOS; IHPS1; NC-NOS; N-NOS; bNOS; Neuronal NOS; Constitutive NOS; NOS type I; Nitric Ocide Synthase; Peptidyl-cysteine S-nitrosylase NOS1. Restriction: For Research Use Only.
Catalog No. 50-228-6270