Learn More
Biomatik Corporation Human Glypican 4 (GPC4) ELISA Kit, 96 tests
Catalog No. 501502642
Encompass_Preferred
The Human Glypican 4 (GPC4) ELISA Kit allows for quantitative determination of Glypican 4 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 31.2-2,000 pg/mL. Sensitivity: 13.3 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O75487. Cross Activity: No significant cross-reactivity or interference between Glypican 4 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2642 Supplier Biomatik Corporation Supplier No. EKU04546
The Human Glypican 4 (GPC4) ELISA Kit allows for quantitative determination of Glypican 4 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 31.2-2,000 pg/mL. Sensitivity: 13.3 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O75487. Cross Activity: No significant cross-reactivity or interference between Glypican 4 and analogues was observed. Restriction: For Research Use Only.