Learn More
Biomatik Corporation Human Golgi Protein 73 (GP73) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04549
Encompass_Preferred
The Human Golgi Protein 73 (GP73) ELISA Kit allows for quantitative determination of Golgi Protein 73 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.625-40 ng/mL. Sensitivity: 0.229 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q8NBJ4. Cross Activity: No significant cross-reactivity or interference between Golgi Protein 73 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2644