Learn More
Biomatik Corporation Mouse Golgi Protein 73 (GP73) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04550
Encompass_Preferred
Mouse Golgi Phosphoprotein 2 (GOLPH2) ELISA Kit for detecting GOLPH2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.94-60ng/ml. Sensitivity: 0.37ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q91XA2. Gene ID: 105348. Target Synonyms: GOLM1; GP73; C9orf155; Golgi Membrane Protein 1; Golgi Protein 73. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2645