Learn More
Biomatik Corporation Human Granulocyte Chemotactic Protein 2 (GCP2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04558
Encompass_Preferred
The Human Granulocyte Chemotactic Protein 2 (GCP2) ELISA Kit allows for quantitative determination of Granulocyte Chemotactic Protein 2 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1,000 pg/mL. Sensitivity: 6.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P80162. Cross Activity: No significant cross-reactivity or interference between Granulocyte Chemotactic Protein 2 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2646