Learn More
Biomatik Corporation Human Parathyroid Hormone Related Protein (PTHrP) ELISA Kit
Supplier:  Biomatik Corporation EKU06489
Encompass_Preferred
Human Parathyroid Hormone Related Protein (PTHrP) ELISA Kit for detecting PTHrP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P12272. Gene ID: 5744. Target Synonyms: PTHLH; HHM; PTHR; PLP; Osteostatin; Parathyroid Hormone Like Hormone. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6291