Learn More
Biomatik Corporation Mouse Granzyme A (GZMA) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04563
Encompass_Preferred
Mouse Granzyme A (GZMA) ELISA Kit for detecting GZMA in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.12-200pg/ml. Sensitivity: 1.35pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P11032. Gene ID: 14938. Target Synonyms: CTLA3; HFSP; HF; H factor; Granzyme 1; Fragmentin-1; Hanukkah factor; Cytotoxic T-Lymphocyte-Associated Serine Esterase 3; CTL tryptase; Cytotoxic T-lymphocyte proteinase 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2649