Learn More
Biomatik Corporation Human Gremlin 1 (GREM1) ELISA Kit, 96 tests
Catalog No. 501502652
Encompass_Preferred
The Human Gremlin 1 (GREM1) ELISA Kit allows for quantitative determination of Gremlin 1 in Serum, Plasma, Urine, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 125-8,000 pg/mL. Sensitivity: 55 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O60565. Cross Activity: No significant cross-reactivity or interference between Gremlin 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2652 Supplier Biomatik Corporation Supplier No. EKU04572
The Human Gremlin 1 (GREM1) ELISA Kit allows for quantitative determination of Gremlin 1 in Serum, Plasma, Urine, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 125-8,000 pg/mL. Sensitivity: 55 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O60565. Cross Activity: No significant cross-reactivity or interference between Gremlin 1 and analogues was observed. Restriction: For Research Use Only.