Learn More
Biomatik Corporation Mouse Peripheral Myelin Protein 22 (PMP22) ELISA Kit
Supplier:  Biomatik Corporation EKU06542
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Mouse. Detection Range: 0.156-10ng/mL. Sensitivity: 0.055ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P16646. Target Synonyms: DSS; CMT1A; CMT1E; GAS3; HMSNIA; HNPP; Sp110; Growth arrest-specific protein 3. Restriction: For Research Use Only.
Catalog No. 50-228-6303