Learn More
Biomatik Corporation Human Growth Arrest And DNA Damage Inducible Protein Alpha (GADD45a) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04574
Encompass_Preferred
The Human Growth Arrest And DNA Damage Inducible Protein Alpha (GADD45a) ELISA Kit allows for quantitative determination of Growth Arrest And DNA Damage Inducible Protein Alpha in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.059 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P24522. Cross Activity: No significant cross-reactivity or interference between Growth Arrest And DNA Damage Inducible Protein Alpha and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2653