Learn More
Biomatik Corporation Human Growth Associated Protein 43 (GAP43) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04580
Encompass_Preferred
The Human Growth Associated Protein 43 (GAP43) ELISA Kit allows for quantitative determination of Growth Associated Protein 43 in Serum, Plasma, Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.061 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P17677. Cross Activity: No significant cross-reactivity or interference between Growth Associated Protein 43 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2656