Learn More
Biomatik Corporation Mouse Growth Associated Protein 43 (GAP43) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04581
Encompass_Preferred
Mouse Growth Associated Protein 43 (GAP43) ELISA Kit for detecting GAP43 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P06837. Gene ID: 14432. Target Synonyms: F1; B-50; PP46; Neuron Growth-Associated Protein 43; Neuromodulin; Axonal Membrane Protein GAP-43; Protein F1; Calmodulin-Binding Protein P-57; Neural Phosphoprotein B-50. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2657