Learn More
Biomatik Corporation Human Platelet Derived Growth Factor Receptor Beta (PDGFRb) ELISA Kit
Supplier:  Biomatik Corporation EKU06693
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.054ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P09619. Target Synonyms: CD140b; JTK12; PDGF-R-Beta; PDGFR; PDGFR1; CD140 antigen-like family member B; Platelet-derived growth factor receptor 1; Beta-type platelet-derived growth factor receptor. Restriction: For Research Use Only.
Catalog No. 50-228-6317