Learn More
Biomatik Corporation Human Growth Differentiation Factor 11 (GDF11) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04585
Encompass_Preferred
The Human Growth Differentiation Factor 11 (GDF11) ELISA Kit allows for quantitative determination of Growth Differentiation Factor 11 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1000 pg/mL. Sensitivity: 6.2 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O95390. Cross Activity: No significant cross-reactivity or interference between Growth Differentiation Factor 11 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2658