Learn More
Biomatik Corporation Human Protein Disulfide Isomerase A6 (PDIA6) ELISA Kit
Supplier:  Biomatik Corporation EKU06906
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.054ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q15084. Target Synonyms: PDI-A6; TXNDC7; P5; ERp5; Thioredoxin Domain Containing 7(Protein Disulfide Isomerase); Protein Disulfide Isomerase-Associated 6; Endoplasmic reticulum protein 5. Restriction: For Research Use Only.
Catalog No. 50-228-6342