Learn More
Biomatik Corporation Gallus Growth Hormone (GH) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04607
Encompass_Preferred
The Gallus Growth Hormone (GH) ELISA Kit allows for quantitative determination of Growth Hormone in Serum, Plasma and other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.055 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: H6UQ67. Cross Activity: No significant cross-reactivity or interference between Growth Hormone and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2667