Learn More
Biomatik Corporation Human Prothymosin Alpha (PTMa) ELISA Kit
Supplier:  Biomatik Corporation EKU06968
Encompass_Preferred
Human Prothymosin Alpha (PTMa) ELISA Kit for detecting PTMa in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.065ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P06454. Gene ID: 5757. Target Synonyms: TMSA; PTM-A; Pro-Thymosin A; Gene Sequence 28; Thymosin alpha-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6346