Learn More
Biomatik Corporation Human Growth Hormone (GH) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04608
Encompass_Preferred
The Human Growth Hormone (GH) ELISA Kit allows for quantitative determination of Growth Hormone in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.059 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P01241. Cross Activity: No significant cross-reactivity or interference between Growth Hormone and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2668