Learn More
Biomatik Corporation Mouse Growth Hormone (GH) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04609
Encompass_Preferred
Mouse Growth Hormone (GH) ELISA Kit for detecting GH in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78-5,000pg/ml. Sensitivity: 34pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P06880. Gene ID: 14599. Target Synonyms: GH1; GH-N; GHN; hGH-N; Somatotropin; Hygetropin; Jintropin; Kigtropin; Pituitary Growth Hormone; Growth Hormone, Normal. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2669