Learn More
Biomatik Corporation Pig Growth Hormone (GH) ELISA Kit, 96 tests
Catalog No. 501502670
Encompass_Preferred
Pig Growth Hormone (GH) ELISA Kit for detecting GH in serum, plasma, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.312-20ng/ml. Sensitivity: 0.117ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01248. Target Synonyms: GH1; GH-N; GHN; hGH-N; Somatotropin; Hygetropin; Jintropin; Kigtropin; Pituitary Growth Hormone; Growth Hormone, Normal. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2670 Supplier Biomatik Corporation Supplier No. EKU04610
Pig Growth Hormone (GH) ELISA Kit for detecting GH in serum, plasma, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.312-20ng/ml. Sensitivity: 0.117ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01248. Target Synonyms: GH1; GH-N; GHN; hGH-N; Somatotropin; Hygetropin; Jintropin; Kigtropin; Pituitary Growth Hormone; Growth Hormone, Normal. Restrictions: For Research Use Only. Not for use in diagnostic procedures.