Learn More
Biomatik Corporation Rat Growth Hormone (GH) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04611
Encompass_Preferred
Rat Growth Hormone (GH) ELISA Kit for detecting GH in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01244. Gene ID: 24391. Target Synonyms: GH1; GH-N; GHN; hGH-N; Somatotropin; Hygetropin; Jintropin; Kigtropin; Pituitary Growth Hormone; Growth Hormone, Normal. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2671