Learn More
Biomatik Corporation Rat Growth Hormone Releasing Hormone (GHRH) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04620
Encompass_Preferred
Rat Growth Hormone Releasing Hormone (GHRH) ELISA Kit for detecting GHRH in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 12.35-1,000pg/ml. Sensitivity: 4.34pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P09916. Gene ID: 29446. Target Synonyms: GRF; GHRF; Somatocrinin; Growth-Hormone-Releasing Factor; Sermorelin; Somatoliberin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2675