Learn More
Biomatik Corporation Human Growth Hormone Secretagogue Receptor (GHSR) ELISA Kit, 96 tests
Catalog No. 501502676
Encompass_Preferred
The Human Growth Hormone Secretagogue Receptor (GHSR) ELISA Kit allows for quantitative determination of Growth Hormone Secretagogue Receptor in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.055 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q92847. Cross Activity: No significant cross-reactivity or interference between Growth Hormone Secretagogue Receptor and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2676 Supplier Biomatik Corporation Supplier No. EKU04621
The Human Growth Hormone Secretagogue Receptor (GHSR) ELISA Kit allows for quantitative determination of Growth Hormone Secretagogue Receptor in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.055 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q92847. Cross Activity: No significant cross-reactivity or interference between Growth Hormone Secretagogue Receptor and analogues was observed. Restriction: For Research Use Only.