Learn More
Biomatik Corporation Human Growth Regulated Oncogene Beta (GROb) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04622
Encompass_Preferred
The Human Growth Regulated Oncogene Beta (GROb) ELISA Kit allows for quantitative determination of Growth Regulated Oncogene Beta in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1000 pg/mL. Sensitivity: 6.3 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P19875. Cross Activity: No significant cross-reactivity or interference between Growth Regulated Oncogene Beta and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2677