Learn More
Biomatik Corporation Sphingomyelin (SPH) ELISA Kit
Supplier:  Biomatik Corporation EKU08699
Encompass_Preferred
Sphingomyelin (SPH) ELISA Kit for detecting SPH in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Universal. Detection Range: 1.23-100ng/ml. Sensitivity: 0.55ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: SM. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6527