Learn More
Biomatik Corporation Human Guanylate Cyclase Activator 2B (GUCA2B) ELISA Kit, 96 tests
Catalog No. 501502680
Encompass_Preferred
The Human Guanylate Cyclase Activator 2B (GUCA2B) ELISA Kit allows for quantitative determination of Guanylate Cyclase Activator 2B in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1,000 pg/mL. Sensitivity: 5.6 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q16661. Cross Activity: No significant cross-reactivity or interference between Guanylate Cyclase Activator 2B and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2680 Supplier Biomatik Corporation Supplier No. EKU04631
The Human Guanylate Cyclase Activator 2B (GUCA2B) ELISA Kit allows for quantitative determination of Guanylate Cyclase Activator 2B in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1,000 pg/mL. Sensitivity: 5.6 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q16661. Cross Activity: No significant cross-reactivity or interference between Guanylate Cyclase Activator 2B and analogues was observed. Restriction: For Research Use Only.