Learn More
Biomatik Corporation Human Haptoglobin (Hpt) ELISA Kit, 96 tests
Catalog No. 501502681
Encompass_Preferred
The Human Haptoglobin (Hpt) ELISA Kit allows for quantitative determination of Haptoglobin in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 6.25-400 ng/mL. Sensitivity: 2.87 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P00738. Cross Activity: No significant cross-reactivity or interference between Haptoglobin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2681 Supplier Biomatik Corporation Supplier No. EKU04634
The Human Haptoglobin (Hpt) ELISA Kit allows for quantitative determination of Haptoglobin in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 6.25-400 ng/mL. Sensitivity: 2.87 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P00738. Cross Activity: No significant cross-reactivity or interference between Haptoglobin and analogues was observed. Restriction: For Research Use Only.