Learn More
Biomatik Corporation Chicken Tumor Necrosis Factor Alpha (TNFa) ELISA Kit
Supplier:  Biomatik Corporation EKU08983
Encompass_Preferred
Chicken Tumor Necrosis Factor Alpha (TNFa) ELISA Kit for detecting TNFa in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 7.8-500pg/ml. Sensitivity: 3.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: AEW10516.1. Target Synonyms: DIF; TNF-A; TNFSF2; Cachectin; Tumor Necrosis Factor Ligand Superfamily Member 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6558